missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Sorbitol hexaacetate, 97%

$100.70 - $337.73

Chemical Identifiers

CAS 7208-47-1
Molecular Formula C18H26O12
Molecular Weight (g/mol) 434.394
MDL Number MFCD00008715
InChI Key NJVBTKVPPOFGAT-XMTFNYHQSA-N
Synonym d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, d-sorbitol hexaacetate, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
PubChem CID 9802952
IUPAC Name [(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
SMILES CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6612114 View Documents Alfa Aesar™
J6612114
25g Each for $100.70
 
AAJ6612122 View Documents Alfa Aesar™
J6612122
100g Each for $337.73
 
Specifications

Chemical Identifiers

7208-47-1
434.394
NJVBTKVPPOFGAT-XMTFNYHQSA-N
9802952
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
C18H26O12
MFCD00008715
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, d-sorbitol hexaacetate, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate

Specifications

White
25g
C18H26O12
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, d-sorbitol hexaacetate, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
434.394
434.39
97%
100°C to 104°C
7208-47-1
MFCD00008715
NJVBTKVPPOFGAT-XMTFNYHQSA-N
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate
9802952
Powder
D-Sorbitol hexaacetate
Safety and Handling

Safety and Handling

TSCA : TSCA