missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Sorbitol hexaacetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J6612114
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-Sorbitol hexaacetate | |
100°C to 104°C | |
C18H26O12 | |
25 g | |
NJVBTKVPPOFGAT-XMTFNYHQSA-N | |
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate | |
9802952 | |
97% |
7208-47-1 | |
White | |
MFCD00008715 | |
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate | |
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
434.394 | |
434.39 | |
Powder |
Chemical Identifiers
7208-47-1 | |
434.394 | |
NJVBTKVPPOFGAT-XMTFNYHQSA-N | |
9802952 | |
CC(=O)OCC(C(C(C(COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
C18H26O12 | |
MFCD00008715 | |
d-sorbitol hexaacetate, sorbitol hexaacetate, sorbitol, acetylated, 2r,3r,4r,5s-1,2,4,5,6-pentakis acetyloxy hexan-3-yl acetate | |
[(2S,3R,4R,5R)-2,3,4,5,6-pentaacetyloxyhexyl] acetate |
Safety and Handling
EINECSNumber : 230-588-6
TSCA : Yes
Recommended Storage : Ambient temperatures