missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Sorbitol, >98%, MP Biomedicals™

$187.29 - $187.29

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym --sorbitol, d---sorbitol, d-glucitol, d-sorbitol, diakarmon, glucarine, glucitol, l-gulitol, sorbitol, sorbol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10293805
View Documents
Mp Biomedicals
0210293805
5kg Each for $187.29
 
Specifications

Chemical Identifiers

50-70-4
182.172
--sorbitol, d---sorbitol, d-glucitol, d-sorbitol, diakarmon, glucarine, glucitol, l-gulitol, sorbitol, sorbol
CHEBI:17924
C(C(C(C(C(CO)O)O)O)O)O
C6H14O6
FBPFZTCFMRRESA-JGWLITMVSA-N
5780
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol

Specifications

1.47g/mL at −5°C
5kg
C6H14O6
FBPFZTCFMRRESA-JGWLITMVSA-N
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
5780
182.2
>98%
White
50-70-4
--sorbitol, d---sorbitol, d-glucitol, d-sorbitol, diakarmon, glucarine, glucitol, l-gulitol, sorbitol, sorbol
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:17924
Crystalline Powder
D-Sorbitol