missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Sorbitol, >98%, MP Biomedicals™

Manufacturer:  Mp Biomedicals 0210293805

 View more versions of this product

Catalog No. ICN10293805



Specifications

Specifications

D-Sorbitol
White
50-70-4
--sorbitol, d---sorbitol, d-glucitol, d-sorbitol, diakarmon, glucarine, glucitol, l-gulitol, sorbitol, sorbol
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:17924
Crystalline Powder
1.47g/mL at −5°C
5kg
C6H14O6
FBPFZTCFMRRESA-JGWLITMVSA-N
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
5780
182.2
>98%

Chemical Identifiers

50-70-4
182.172
--sorbitol, d---sorbitol, d-glucitol, d-sorbitol, diakarmon, glucarine, glucitol, l-gulitol, sorbitol, sorbol
CHEBI:17924
C(C(C(C(C(CO)O)O)O)O)O
C6H14O6
FBPFZTCFMRRESA-JGWLITMVSA-N
5780
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
Documents