missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Serine methyleester hydrochloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC396790050

 View more versions of this product

Catalog No. AC396790050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5% (Argentometry)
Liquid
Clear colorless to yellow
−2.5° to −4.5° (20°C, 589nm) (C=4, methanol)
5874-57-7
Glass bottle
D-Serine methyleester hydrochloride
155.58
5g
C4H9NO3·ClH
Authentic
MFCD00066121
HOCH2CH(NH2)CO2CH3·HCl
98%

Chemical Identifiers

98%
HOCH2CH(NH2)CO2CH3·HCl
MFCD00066121
≥97.5% (Argentometry)
C4H9NO3·ClH
155.58
D-Serine methyleester hydrochloride