Please login to your online account to display your discounted pricing

D-Serine methyleester hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material D-Serine methyleester hydrochloride
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C4H9NO3·ClH
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Formula Weight 155.58
MDL Number MFCD00066121
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC396790050

  SDS
thermo fisher scientific
AC396790050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC396790250

  SDS
fisher scientific acros
AC396790250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (Argentometry)
Formula Weight 155.58
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
Molecular Formula C4H9NO3·ClH
Specific Rotation −2.5° to −4.5° (20°C, 589nm) (C=4, methanol)
Infrared Spectrum Authentic
CAS 5874-57-7
MDL Number MFCD00066121
Packaging Glass bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Chemical Name or Material D-Serine methyleester hydrochloride
Assay Percent Range 98%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.