missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Serine benzyl ester hydrochloride, 95%

$51.35 - $209.73

Chemical Identifiers

CAS 151651-44-4
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.676
MDL Number MFCD00191017
InChI Key MGZWCDQAKCHOBX-SBSPUUFOSA-N
Synonym h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl
PubChem CID 11776084
IUPAC Name benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)C(CO)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6367803 View Documents Alfa Aesar™
H6367803
1g Each for $51.35
 
AAH6367806 View Documents Alfa Aesar™
H6367806
5g Each for $209.73
 
Specifications

Chemical Identifiers

151651-44-4
231.676
MGZWCDQAKCHOBX-SBSPUUFOSA-N
11776084
C1=CC=C(C=C1)COC(=O)C(CO)N.Cl
C10H14ClNO3
MFCD00191017
h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl
benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride

Specifications

151651-44-4
MFCD00191017
MGZWCDQAKCHOBX-SBSPUUFOSA-N
benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
11776084
Solid
169°C to 174°C
D-Serine benzyl ester hydrochloride
C10H14ClNO3
h-d-ser-obzl.hcl, h-d-ser-obzl hcl, d-serine benzyl ester hydrochloride, r-benzyl 2-amino-3-hydroxypropanoate hydrochloride, d-serinebenzylesterhydrochloride, benzyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-serine-obzl hcl, h-d-ser-obzlhcl, h-d-ser-obzl?cl, h-d-ser-obzl. hcl
C1=CC=C(C=C1)COC(=O)C(CO)N.Cl
231.676
231.68
95%
1g
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C