missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(-)-Salicin, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132590050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D(-)-Salicin | |
138-52-3 | |
White | |
99% min. (HPLC) | |
C13H18O7 | |
5 g | |
15, 8460 | |
-61.5 | |
Solubility in water: 36g/L (15°C),250g/L (60°C). Other solubilities: insoluble in ether | |
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O | |
286.27 | |
CHEBI:17814 | |
99+% |
99+% | |
197°C to 200°C | |
Authentic | |
Glass bottle | |
MFCD00006590 | |
17, V,7, 113 | |
−61.50° (20°C c=5,water) | |
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
NGFMICBWJRZIBI-UJPOAAIJSA-N | |
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol | |
439503 | |
286.27 | |
Fine Crystalline Powder |
Chemical Identifiers
138-52-3 | |
286.27 | |
NGFMICBWJRZIBI-UJPOAAIJSA-N | |
439503 | |
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
C13H18O7 | |
MFCD00006590 | |
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
CHEBI:17814 | |
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
GHS P Statement
Wear eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 205-331-6
RTECSNumber : LZ5901700
TSCA : TSCA
RUO – Research Use Only