Please login to your online account to display your discounted pricing

D-(-)-Ribose, MP Biomedicals


Quantity 25g

Manufacturer: mp biomedicals  10281925

 View more versions of this product

Catalog No. ICN-10281925

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Color White
Quantity 25g
EINECS Number 200-059-4
Name Note Cell Biology and Analysis
Assay Percent Range ≥97%
Molecular Formula C5H10O5
MDL Number MFCD00135453
CAS 50-69-1
Chemical Name or Material D-(-)-Ribose
Synonym D-Ribofuranose
Recommended Storage Store at +4°C.

Specifications

Chemical Name or Material D-(-)-Ribose
Name Note Cell Biology and Analysis
CAS 50-69-1
Physical Form Powder
Assay Percent Range ≥97%
Molecular Formula C5H10O5
MDL Number MFCD00135453
Synonym D-Ribofuranose
Accessories

Store at +4°C.