Please login to your online account to display your discounted pricing

D-(-)-Ribose, MP Biomedicals

$54.72 - $54.72

Chemical Identifiers

Chemical Name or Material D-(-)-Ribose
Name Note Cell Biology and Analysis
CAS 50-69-1
Physical Form Powder
Assay Percent Range ≥97%
Molecular Formula C5H10O5
MDL Number MFCD00135453
Synonym D-Ribofuranose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN-10281925

 
mp biomedicals
10281925
25g Each for $54.72
Description & Specifications

Specifications

Physical Form Powder
Color White
Quantity 25g
EINECS Number 200-059-4
Name Note Cell Biology and Analysis
Assay Percent Range ≥97%
Molecular Formula C5H10O5
MDL Number MFCD00135453
CAS 50-69-1
Chemical Name or Material D-(-)-Ribose
Synonym D-Ribofuranose
Recommended Storage Store at +4°C.