missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Ribono-1,4-lactone 97.0+%, TCI America™

Manufacturer:  TCI America R00635G

 View more versions of this product

Catalog No. R00635G



Specifications

Specifications

D-(+)-Ribono-1,4-lactone
86°C
5336-08-3
MFCD00063241
CUOKHACJLGPRHD-UHFFFAOYNA-N
(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
111064
148.11
≥97.0% (GC)
Undesignated
5g
C5H8O5
ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
C(C1C(C(C(=O)O1)O)O)O
148.11
CHEBI:74168
Crystal-Powder at 20°C

Chemical Identifiers

5336-08-3
148.11
CUOKHACJLGPRHD-UHFFFAOYNA-N
111064
(3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one
C5H8O5
MFCD00063241
ribonolactone, d-ribono-1,4-lactone, d-ribonolactone, ribono-gamma-lactone, d +-ribonic acid gamma-lactone, d-+-ribono-1,4-lactone, d-ribopentono-1,4-lactone, d-ribono-gamma-lactone, d-+-ribonic acid gamma-lactone, d-ribonic acid-1,4-lactone
CHEBI:74168
C(C1C(C(C(=O)O1)O)O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents