Please login to your online account to display your discounted pricing

β-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material β-D-Ribofuranose 1,2,3,5-tetraacetate
CAS 13035-61-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0% (on dry substance)
Assay Percent Range 98+%
Molecular Formula C13H18O9
Formula Weight 318.27
MDL Number MFCD00005358
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170121000

  SDS
thermo fisher scientific
AC170121000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC170120250

  SDS
fisher scientific acros
170120250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0% (on dry substance)
Formula Weight 318.27
Physical Form Crystalline Powder
Quantity 100g
Color White to almost white
Specific Rotation Condition −15.40° (20°C c=7,MeOH)
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C13H18O9
Residue after Ignition 0.1% max.
Heavy Metals (as Pb) 20ppm max.
Chemical Name or Material β-D-Ribofuranose 1,2,3,5-tetraacetate
Loss on Drying 1% max. (60°C, 2 hrs) (vacuum)
Specific Rotation On dry substance: -14.9° to -15.9° (20°C, 589nm) (c=7, MeOH)
CAS 13035-61-5
Assay Percent Range 98+%
MDL Number MFCD00005358