missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Psicose, TCI America™

Manufacturer:  TCI America P16991G

 View more versions of this product

Catalog No. P16991G



Specifications

Specifications

D-Psicose
≥99.0% (HPLC)
MFCD00083478
White
BJHIKXHVCXFQLS-PUFIMZNGSA-N
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
90008
180.16
551-68-8
C6H12O6
109°C
1g
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:27605
Crystal-Powder at 20°C

Chemical Identifiers

551-68-8
180.156
BJHIKXHVCXFQLS-PUFIMZNGSA-N
CHEBI:27605
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
MFCD00083478
90008
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Safety and Handling

Safety and Handling

TSCA : No

Documents