missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Psicose, TCI America™

Manufacturer:  TCI America P1699100MG

 View more versions of this product

Catalog No. P1699100MG



Specifications

Specifications

D-Psicose
109°C
551-68-8
MFCD00083478
BJHIKXHVCXFQLS-PUFIMZNGSA-N
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
90008
180.16
≥99.0% (HPLC)
White
100mg
C6H12O6
erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:27605
Crystal-Powder at 20°C

Chemical Identifiers

551-68-8
180.156
BJHIKXHVCXFQLS-PUFIMZNGSA-N
90008
(3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
MFCD00083478
erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
CHEBI:27605
C(C(C(C(C(=O)CO)O)O)O)O
Safety and Handling

Safety and Handling

TSCA : No

Documents