missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Propargylglycine, 97%

$97.92 - $428.70

Chemical Identifiers

CAS 23235-03-2
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.116
MDL Number MFCD00237407
InChI Key DGYHPLMPMRKMPD-SCSAIBSYSA-N
Synonym d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
PubChem CID 6992713
IUPAC Name (2R)-2-aminopent-4-ynoic acid
SMILES C#CCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6579803 View Documents Alfa Aesar™
H6579803
1g Each for $97.92
 
AAH6579806 View Documents Alfa Aesar™
H6579806
5g Each for $428.70
 
Description

Description

Applications
A useful biochemical for proteomics research. Also been used for affinity labeling of hog kidney D-amino acid oxidase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

23235-03-2
113.116
DGYHPLMPMRKMPD-SCSAIBSYSA-N
6992713
C#CCC(C(=O)O)N
C5H7NO2
MFCD00237407
d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
(2R)-2-aminopent-4-ynoic acid

Specifications

23235-03-2
MFCD00237407
DGYHPLMPMRKMPD-SCSAIBSYSA-N
(2R)-2-aminopent-4-ynoic acid
6992713
Powder
White
1g
D-Propargylglycine
C5H7NO2
d-propargylglycine, r-2-aminopent-4-ynoic acid, r-alpha-propargylglycine, unii-2252qef2pl, 4-pentynoic acid, 2-amino-, 2r, 2r-2-aminopent-4-ynoic acid, r-2-propargylglycine, r-2-amino-4-pentynoic acid, d-2-propynylglycine, propargylglycine, d
C#CCC(C(=O)O)N
113.116
113.12
97%
205°C (decomposition)
Odorless
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning