missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Proline tert-Butyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America P17281G

 View more versions of this product

Catalog No. P17281G



Specifications

Specifications

D-Proline tert-Butyl Ester Hydrochloride
≥98.0% (GC,T)
MFCD00153460
IUUYANMOEMBTBV-OGFXRTJISA-N
tert-butyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
44629909
Crystal-Powder at 20°C
94°C
184719-80-0
C9H18ClNO2
H-D-Pro-OtBu.HCl
CC(C)(C)OC(=O)C1CCCN1.Cl
207.698
207.70
White
1g

Chemical Identifiers

184719-80-0
207.698
IUUYANMOEMBTBV-OGFXRTJISA-N
44629909
CC(C)(C)OC(=O)C1CCCN1.Cl
C9H18ClNO2
MFCD00153460
H-D-Pro-OtBu.HCl
tert-butyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents