missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Proline methyl ester hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445590010
Specifications
D-Proline methyl ester hydrochloride | |
Glass bottle | |
White | |
1g | |
97.5 to max. 102.5% min. (Argentometry) | |
d-proline methyl ester hydrochloride, d-proline, methyl ester, hydrochloride, h-d-pro-ome hcl, h-d-pro-ome.hcl, methyl 2r-pyrrolidine-2-carboxylate hydrochloride, methyl d-prolinate hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, r-methyl pyrrolidine-2-carboxylate hydrochloride, r-proline methyl ester hydrochloride, unii-c15epu1m46 | |
COC(=O)C1CCCN1.Cl | |
165.62 | |
165.62 | |
98% |
Authentic | |
35 | |
42°C to 49°C | |
65365-28-8 | |
C6H11NO2·ClH | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
44629857 | |
Powder or Crystalline Powder |
Chemical Identifiers
65365-28-8 | |
165.62 | |
d-proline methyl ester hydrochloride, d-proline, methyl ester, hydrochloride, h-d-pro-ome hcl, h-d-pro-ome.hcl, methyl 2r-pyrrolidine-2-carboxylate hydrochloride, methyl d-prolinate hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, r-methyl pyrrolidine-2-carboxylate hydrochloride, r-proline methyl ester hydrochloride, unii-c15epu1m46 | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
C6H11NO2·ClH | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
44629857 | |
COC(=O)C1CCCN1.Cl |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning