missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Proline ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6491014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-Proline ethyl ester hydrochloride | |
Yellow | |
MFCD00191003 | |
r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl | |
CCOC(=O)C1CCCN1.Cl | |
179.644 | |
179.64 | |
25 g |
131477-20-8 | |
C7H14ClNO2 | |
7170607 | |
DUJGQVVONTYHLT-FYZOBXCZSA-N | |
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
56777374 | |
98% | |
Liquid |
Chemical Identifiers
131477-20-8 | |
179.644 | |
DUJGQVVONTYHLT-FYZOBXCZSA-N | |
56777374 | |
CCOC(=O)C1CCCN1.Cl |
C7H14ClNO2 | |
MFCD00191003 | |
r-ethyl pyrrolidine-2-carboxylate hydrochloride, d-proline ethyl ester hydrochloride, h-d-pro-oet hcl, h-d-pro-oet hydrochloride, d-proline, ethyl ester, hydrochloride, ethyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-oet.hcl, h-d-pro-oet . hci, h-d-pro-oet?cl, d-proline ethyl ester hcl | |
ethyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only