missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Proline Benzyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America P172625G

 View more versions of this product

Catalog No. P172625G



Specifications

Specifications

D-Proline Benzyl Ester Hydrochloride
≥98.0% (HPLC,N)
MFCD00153458
NEDMOHHWRPHBAL-RFVHGSKJSA-N
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
22827650
Crystal-Powder at 20°C
25g
53843-90-6
C12H16ClNO2
H-D-Pro-OBzl.HCl
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
241.715
241.72
White

Chemical Identifiers

53843-90-6
241.715
NEDMOHHWRPHBAL-RFVHGSKJSA-N
22827650
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
C12H16ClNO2
MFCD00153458
H-D-Pro-OBzl.HCl
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents