missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Proline benzyl ester hydrochloride, 97%

$69.70 - $316.14

Chemical Identifiers

CAS 53843-90-6
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
MDL Number MFCD00153458
InChI Key NEDMOHHWRPHBAL-RFVHGSKJSA-N
Synonym h-d-pro-obzl hcl, h-d-pro-obzl.hcl, d-proline benzyl ester hydrochloride, d-proline, phenylmethyl ester, hydrochloride, r-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-obzl-hcl, h-d-pro-obzl inverted exclamation mark currencyhcl, d-proline benzylester hydrochloride, d-proline-benzyl-ester hydrochloride
PubChem CID 22827650
IUPAC Name benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6580403 View Documents Alfa Aesar™
H6580403
1g Each for $69.70
 
AAH6580406 View Documents Alfa Aesar™
H6580406
5g Each for $316.14
 
Specifications

Chemical Identifiers

53843-90-6
241.715
NEDMOHHWRPHBAL-RFVHGSKJSA-N
22827650
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
C12H16ClNO2
MFCD00153458
h-d-pro-obzl hcl, h-d-pro-obzl.hcl, d-proline benzyl ester hydrochloride, d-proline, phenylmethyl ester, hydrochloride, r-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-obzl-hcl, h-d-pro-obzl inverted exclamation mark currencyhcl, d-proline benzylester hydrochloride, d-proline-benzyl-ester hydrochloride
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride

Specifications

53843-90-6
MFCD00153458
h-d-pro-obzl hcl, h-d-pro-obzl.hcl, d-proline benzyl ester hydrochloride, d-proline, phenylmethyl ester, hydrochloride, r-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl 2r-pyrrolidine-2-carboxylate hydrochloride, h-d-pro-obzl-hcl, h-d-pro-obzl inverted exclamation mark currencyhcl, d-proline benzylester hydrochloride, d-proline-benzyl-ester hydrochloride
C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl
241.715
241.71
97%
D-Proline benzyl ester hydrochloride
C12H16ClNO2
6852422
NEDMOHHWRPHBAL-RFVHGSKJSA-N
benzyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
22827650
Solid
1g