missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Prolinamide hydrochloride, 97%

$122.45 - $487.31

Chemical Identifiers

CAS 50894-62-7
Molecular Formula C5H11ClN2O
Molecular Weight (g/mol) 150.606
MDL Number MFCD00237235
InChI Key CSKSDAVTCKIENY-PGMHMLKASA-N
Synonym r-pyrrolidine-2-carboxamide hydrochloride, h-d-pro-nh2 hcl, d-prolinamide hydrochloride, d-proline amide hydrochloride, d-prolinamide hcl, d-prolinamide hydrochloride, r-prolinamide hydrochloride, 2r-pyrrolidine-2-carboxamide hydrochloride, d-prolin amide hydrochloride, h-d-pro-nh hcl
PubChem CID 46735192
IUPAC Name (2R)-pyrrolidine-2-carboxamide;hydrochloride
SMILES C1CC(NC1)C(=O)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6569006 View Documents Alfa Aesar™
H6569006
5g Each for $122.45
 
AAH6569014 View Documents Alfa Aesar™
H6569014
25g Each for $487.31
 
Specifications

Chemical Identifiers

50894-62-7
150.606
CSKSDAVTCKIENY-PGMHMLKASA-N
46735192
C1CC(NC1)C(=O)N.Cl
C5H11ClN2O
MFCD00237235
r-pyrrolidine-2-carboxamide hydrochloride, h-d-pro-nh2 hcl, d-prolinamide hydrochloride, d-proline amide hydrochloride, d-prolinamide hcl, d-prolinamide hydrochloride, r-prolinamide hydrochloride, 2r-pyrrolidine-2-carboxamide hydrochloride, d-prolin amide hydrochloride, h-d-pro-nh hcl
(2R)-pyrrolidine-2-carboxamide;hydrochloride

Specifications

50894-62-7
MFCD00237235
r-pyrrolidine-2-carboxamide hydrochloride, h-d-pro-nh2 hcl, d-prolinamide hydrochloride, d-proline amide hydrochloride, d-prolinamide hcl, d-prolinamide hydrochloride, r-prolinamide hydrochloride, 2r-pyrrolidine-2-carboxamide hydrochloride, d-prolin amide hydrochloride, h-d-pro-nh hcl
C1CC(NC1)C(=O)N.Cl
150.606
150.61
97%
D-Prolinamide hydrochloride
C5H11ClN2O
6280978
CSKSDAVTCKIENY-PGMHMLKASA-N
(2R)-pyrrolidine-2-carboxamide;hydrochloride
46735192
Solid
5g