Please login to your online account to display your discounted pricing

Alfa Aesar™ D-(-)-Prolinamide, 98%

$83.49 - $307.38

Chemical Identifiers

Chemical Name or Material D-(-)-Prolinamide
CAS 62937-45-5
Molecular Formula C5H10N2O
Formula Weight 114.15
MDL Number MFCD00153457
Synonym H-D-Pro-NH2; (R)-Pyrrolidine-2-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3090103

  SDS
alfa aesar™
H3090103
1g Each for $83.49

AAH3090106

  SDS
alfa aesar™
H3090106
5g Each for $307.38
Description & Specifications

Specifications

Formula Weight 114.15
Synonym H-D-Pro-NH2; (R)-Pyrrolidine-2-carboxamide
Melting Point 100° to 103°C
Quantity 1g
CAS 62937-45-5
Assay 98%
Chemical Name or Material D-(-)-Prolinamide
Optical Rotation +107° (c=2 in Ethanol)
Molecular Formula C5H10N2O
MDL Number MFCD00153457

Applications
It can be used in the synthesis of proline derivatives as efficient organocatalysts for aldol reaction on water.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.