missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(-)-Phenylglycinamide, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 346700250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D(-)-Phenylglycinamide | |
6485-67-2 | |
98.5% min. (GC) | |
C8H10N2O | |
MFCD06799064 | |
−48.5° (20°C c=0.55,EtOH) | |
r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide | |
KIYRSYYOVDHSPG-SSDOTTSWSA-N | |
(2R)-2-amino-2-phenylacetamide | |
729987 | |
99% |
99% | |
Authentic | |
Glass bottle | |
C6H5CH(NH2)C(O)NH2 | |
25g | |
− 48.50 | |
Solubility in water: soluble. Other solubilities: better soluble ph <6 | |
C1=CC=C(C=C1)C(C(=O)N)N | |
150.18 | |
150.18 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO â Research Use Only