missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Fisher Scientific 442620250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-Phenylalanine methyl ester hydrochloride | |
White | |
97.5% min. (HPLC) | |
Authentic | |
Glass bottle | |
d-phenylalanine methyl ester hcl, d-phenylalanine methyl ester hydrochloride, d-phenylalanine, methyl ester, hydrochloride, h-d-phe-ome hcl, h-d-phe-ome.hcl, methyl 2r-2-amino-3-phenylpropanoate hydrochloride, methyl d-phenylalaninate hydrochloride, r-methyl 2-amino-3-phenylpropanoate hydrochloride, r-phenylalanine methyl ester hydrochloride | |
Cl.COC(=O)C(N)CC1=CC=CC=C1 | |
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride | |
11481330 | |
Powder or Crystalline Powder |
13033-84-6 | |
157°C to 159°C | |
C10H14ClNO2 | |
MFCD00066112 | |
25g | |
SWVMLNPDTIFDDY-UHFFFAOYNA-N | |
-35.76 | |
215.68 | |
215.68 | |
98% |
RUO – Research Use Only