missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Phenylalanine Methyl Ester Hydrochloride, TCI America™

Manufacturer:  TCI America P172525G

 View more versions of this product

Catalog No. P172525G



Specifications

Specifications

D-Phenylalanine Methyl Ester Hydrochloride
160°C
13033-84-6
MFCD00066112
SWVMLNPDTIFDDY-SBSPUUFOSA-N
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
11481330
Crystal-Powder at 20°C
White
25g
C10H14ClNO2
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, d-phenylalanine methyl ester hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
COC(=O)C(CC1=CC=CC=C1)N.Cl
215.677
215.68
≥98.0% (HPLC,T)

Chemical Identifiers

13033-84-6
215.677
SWVMLNPDTIFDDY-SBSPUUFOSA-N
11481330
COC(=O)C(CC1=CC=CC=C1)N.Cl
C10H14ClNO2
MFCD00066112
d-phenylalanine methyl ester hydrochloride, h-d-phe-ome.hcl, d-phenylalanine methyl ester hcl, h-d-phe-ome hcl, r-methyl 2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine, methyl ester, hydrochloride, methyl d-phenylalaninate hydrochloride, d-phenylalanine methyl ester hydrochloride, r-phenylalanine methyl ester hydrochloride, methyl 2r-2-amino-3-phenylpropanoate hydrochloride
methyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents