Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Phenylalanine benzyl ester p-toluenesulfonate, 98%

Chemical Identifiers

Chemical Name or Material D-Phenylalanine benzyl ester p-toluenesulfonate
CAS 28607-46-7
Formula Weight 427.51
MDL Number MFCD00066129
Beilstein 5375462
Synonym H-D-Phe-OBzl.Tos; D-Phenylalanine benzyl ester tosylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6549606

 
alfa aesar™
H6549606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6549614

 
alfa aesar™
H6549614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6549622

 
alfa aesar™
H6549622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 427.51
CAS 28607-46-7
Chemical Name or Material D-Phenylalanine benzyl ester p-toluenesulfonate
Assay 98%
Synonym H-D-Phe-OBzl.Tos; D-Phenylalanine benzyl ester tosylate
Physical Form Solid
Beilstein 5375462
Quantity 5g
Storage Note 1 2° to 8°C
MDL Number MFCD00066129
Safety and Handling

2° to 8°C