missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Phenylalaninamide hydrochloride, 98%

$77.88 - $1,028.59

Chemical Identifiers

CAS 71666-94-9
Molecular Formula C9H13ClN2O
Molecular Weight (g/mol) 200.666
MDL Number MFCD00153454
InChI Key KLHLGTPNBQXSJT-DDWIOCJRSA-N
Synonym d-phenylalanine amide hydrochloride, h-d-phe-nh2 hcl, 2r-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, h-d-phe-nh2.hcl, h-d-phe-nh2?cl, d-phe-nh2? currencyhcl, d-phenylalaninamide hydrochloride, d-phe-nh2? inverted exclamation markehcl, 2r-2-amino-3-phenylpropionamide hydrochloride
PubChem CID 56777394
IUPAC Name (2R)-2-amino-3-phenylpropanamide;hydrochloride
SMILES C1=CC=C(C=C1)CC(C(=O)N)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6585306 View Documents Alfa Aesar™
H6585306
5g Each for $77.88
 
AAH6585314 View Documents Alfa Aesar™
H6585314
25g Each for $308.42
 
AAH6585322 View Documents Alfa Aesar™
H6585322
100g Each for $1,028.59
 
Specifications

Chemical Identifiers

71666-94-9
200.666
KLHLGTPNBQXSJT-DDWIOCJRSA-N
56777394
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
C9H13ClN2O
MFCD00153454
d-phenylalanine amide hydrochloride, h-d-phe-nh2 hcl, 2r-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, h-d-phe-nh2.hcl, h-d-phe-nh2?cl, d-phe-nh2? currencyhcl, d-phenylalaninamide hydrochloride, d-phe-nh2? inverted exclamation markehcl, 2r-2-amino-3-phenylpropionamide hydrochloride
(2R)-2-amino-3-phenylpropanamide;hydrochloride

Specifications

71666-94-9
MFCD00153454
d-phenylalanine amide hydrochloride, h-d-phe-nh2 hcl, 2r-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, h-d-phe-nh2.hcl, h-d-phe-nh2?cl, d-phe-nh2? currencyhcl, d-phenylalaninamide hydrochloride, d-phe-nh2? inverted exclamation markehcl, 2r-2-amino-3-phenylpropionamide hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
200.666
200.67
98%
D-Phenylalaninamide hydrochloride
C9H13ClN2O
3915663
KLHLGTPNBQXSJT-DDWIOCJRSA-N
(2R)-2-amino-3-phenylpropanamide;hydrochloride
56777394
Solid
5g