missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Phenylalaninamide hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6585314

 View more versions of this product

Catalog No. AAH6585314



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-Phenylalaninamide hydrochloride
C9H13ClN2O
3915663
KLHLGTPNBQXSJT-DDWIOCJRSA-N
(2R)-2-amino-3-phenylpropanamide;hydrochloride
56777394
98%
Solid
71666-94-9
MFCD00153454
d-phenylalanine amide hydrochloride, h-d-phe-nh2 hcl, 2r-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, h-d-phe-nh2.hcl, h-d-phe-nh2?cl, d-phe-nh2? currencyhcl, d-phenylalaninamide hydrochloride, d-phe-nh2? inverted exclamation markehcl, 2r-2-amino-3-phenylpropionamide hydrochloride
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
200.666
200.67
25 g

Chemical Identifiers

71666-94-9
200.666
KLHLGTPNBQXSJT-DDWIOCJRSA-N
56777394
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
C9H13ClN2O
MFCD00153454
d-phenylalanine amide hydrochloride, h-d-phe-nh2 hcl, 2r-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, h-d-phe-nh2.hcl, h-d-phe-nh2?cl, d-phe-nh2? currencyhcl, d-phenylalaninamide hydrochloride, d-phe-nh2? inverted exclamation markehcl, 2r-2-amino-3-phenylpropionamide hydrochloride
(2R)-2-amino-3-phenylpropanamide;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only