Please login to your online account to display your discounted pricing

Alfa Aesar™ [D-Phe&superscript_1;&superscript_2;] Bombesin

For cellular and molecular biology applications

$109.43 - $314.01

Specifications

Product Type [D-Phe12] Bombesin
Molecular Weight 1629.88
Storage Requirements -30° to -10°C
Sequence Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-D-Phe-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66244LB0

 
alfa aesar™
J66244LB0
5mg Each for $314.01

AAJ66244MCR

 
alfa aesar™
J66244MCR
1mg Each for $109.43
Description & Specifications

Specifications

Product Type [D-Phe12] Bombesin
Molecular Weight 1629.88
Storage Requirements -30° to -10°C
Sequence Glp-Gln-Arg-Leu-Gly-Asn-Gln-Trp-Ala-Val-Gly-D-Phe-Leu-Met-NH2
Format Solid
Common Name Bombesin Receptor Antagonist
Regulatory Status RUO
Purification Method HPLC