missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(-)-Pantolactone 98.0+%, TCI America™

Manufacturer:  TCI America P0011500G

 View more versions of this product

Catalog No. P0011500G



Specifications

Specifications

D-(-)-Pantolactone
92°C
599-04-2
MFCD00005392
SERHXTVXHNVDKA-BYPYZUCNSA-N
(3R)-3-hydroxy-4,4-dimethyloxolan-2-one
439368
130.14
≥98.0% (T)
White-Yellow
500g
C6H10O3
d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
CC1(COC(=O)C1O)C
130.143
CHEBI:16719
Crystal-Powder at 20°C

Chemical Identifiers

599-04-2
130.143
SERHXTVXHNVDKA-BYPYZUCNSA-N
439368
(3R)-3-hydroxy-4,4-dimethyloxolan-2-one
C6H10O3
MFCD00005392
d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
CHEBI:16719
CC1(COC(=O)C1O)C
Safety and Handling

Safety and Handling

TSCA : Yes

Documents