Please login to your online account to display your discounted pricing

Alfa Aesar™ D-Ornithine hydrochloride, 98+%

$33.76 - $116.92

Chemical Identifiers

Chemical Name or Material D-Ornithine hydrochloride
CAS 16682-12-5
Molecular Formula C5H12N2O2·HCl
Formula Weight 168.62
MDL Number MFCD00012917
Beilstein 4153339
Synonym (R)-(-)-2,5-Diaminopentanoic acid hydrochloride; H-D-Orn-OH·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0079303

  SDS
alfa aesar™
L0079303
1g Each for $33.76

AAL0079306

  SDS
alfa aesar™
L0079306
5g Each for $116.92
Description & Specifications

Specifications

Formula Weight 168.62
CAS 16682-12-5
Chemical Name or Material D-Ornithine hydrochloride
Molecular Formula C5H12N2O2·HCl
MDL Number MFCD00012917
Beilstein 4153339
Synonym (R)-(-)-2,5-Diaminopentanoic acid hydrochloride; H-D-Orn-OH·HCl
Quantity 1g
Assay ≥98%
Melting Point 237°C
Optical Rotation −23° (c=5 in 5N HCl)