missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LaMotte™ D-Net with Expandable Pole

Bag attached to metal frame with clamp rings.

Manufacturer:  LaMotte™ 0167B

 View more versions of this product

Catalog No. S85027B



Description

Description

  • 12 in. (30.5cm) rim dip net
  • Rim fitted with heavy canvas bag, approximately 6.5 in. deep (16.5cm)
  • Bag bottom made of approximately 500μm nylon mesh
  • Canvas skirt extends 3 in. (7.6cm) below bag bottom to protect mesh
  • Aluminum pole extends from 52 to 84 in. (137.2 to 213.3cm)

Specifications

Specifications

Replacement bag
D-net
Canvas, Nylon
Environmental education
SDS
Documents
Product Certifications
Promotions

Promotions