Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-4-phosphate ammonium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-4-phosphate ammonium salt
CAS 142760-33-6
Formula Weight 276.16
MDL Number MFCD00135425
Synonym Ins(4)P1 ammonium salt; 4-IP1 ammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67010EXZ

 
alfa aesar™
J67010EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.16
CAS 142760-33-6
MDL Number MFCD00135425
Chemical Name or Material D-myo-Inositol-4-phosphate ammonium salt
Physical Form Lyophilized Powder
Synonym Ins(4)P1 ammonium salt; 4-IP1 ammonium salt
Quantity 0.1mg
Storage Note 1 -30° to -10°C
Color White to off-white
Assay 98%
Safety and Handling

-30° to -10°C