Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-4,5-bisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-4,5-bisphosphate sodium salt
Formula Weight 384.08
MDL Number MFCD12912287
Synonym 4,5-IP2 sodium salt; Ins(4,5)-P2 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67071EXD

 
alfa aesar™
J67071EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67071EXZ

 
alfa aesar™
J67071EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67071MCR

 
alfa aesar™
J67071MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 384.08
Chemical Name or Material D-myo-Inositol-4,5-bisphosphate sodium salt
MDL Number MFCD12912287
Assay 98%
Synonym 4,5-IP2 sodium salt; Ins(4,5)-P2 sodium salt
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Quantity 0.5mg
Color White to off-white
Safety and Handling

-30° to -10°C