Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-4,5-bisphosphate sodium salt, 98%


Quantity 0.5mg

Manufacturer: alfa aesar™  J67071EXD

 View more versions of this product

Catalog No. AAJ67071EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 384.08
Chemical Name or Material D-myo-Inositol-4,5-bisphosphate sodium salt
Assay 98%
MDL Number MFCD12912287
Physical Form Lyophilized Powder
Synonym 4,5-IP2 sodium salt; Ins(4,5)-P2 sodium salt
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Color White to off-white

Specifications

Chemical Name or Material D-myo-Inositol-4,5-bisphosphate sodium salt
Formula Weight 384.08
MDL Number MFCD12912287
Synonym 4,5-IP2 sodium salt; Ins(4,5)-P2 sodium salt
Accessories

-30° to -10°C