missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-myo-Inositol-3-phosphate sodium salt, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J67072MCR
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-myo-Inositol-3-phosphate sodium salt | |
1mg | |
Soluble in water | |
98% |
MFCD05864254 | |
3-IP1 sodium salt; Ins(3)P1 sodium salt | |
282.12 |
Chemical Identifiers
MFCD05864254 | |
3-IP1 sodium salt; Ins(3)P1 sodium salt |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO â Research Use Only