missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,5,6-trisphosphate sodium salt, 98%

Chemical Identifiers

MDL Number MFCD26967929
Synonym Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67044EXZ
View Documents
Alfa Aesar™
J67044EXZ
0.1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67044EXD
View Documents
Alfa Aesar™
J67044EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

MFCD26967929
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt

Specifications

White
MFCD26967929
486.04
98%
0.1mg
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt
Lyophilized Powder
D-myo-Inositol-2,5,6-trisphosphate sodium salt
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C