missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,5,6-trisphosphate sodium salt, 98%

Manufacturer:  Alfa Aesar™ J67044EXZ

 View more versions of this product

Catalog No. AAJ67044EXZ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

D-myo-Inositol-2,5,6-trisphosphate sodium salt
0.1mg
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt
Lyophilized Powder
White
MFCD26967929
486.04
98%

Chemical Identifiers

MFCD26967929
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents