missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,4-bisphosphate sodium salt, 98%

Manufacturer:  Alfa Aesar™ J67078MCR

 View more versions of this product

Catalog No. AAJ67078MCR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

White
MFCD18427940
384.08
98%
1mg
Ins(2,4)-P2 sodium salt; 2,4-IP2 sodium salt
Lyophilized Powder

Chemical Identifiers

MFCD18427940
Ins(2,4)-P2 sodium salt; 2,4-IP2 sodium salt
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents