Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,4,5-trisphosphate sodium salt, 98%


Quantity 0.1mg

Manufacturer: alfa aesar™  J67156EXZ

 View more versions of this product

Catalog No. AAJ67156EXZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-2,4,5-trisphosphate sodium salt
MDL Number MFCD05864256
Assay 98%
Color White to off-white
Storage Note 1 -30° to -10°C
Quantity 0.1mg
Physical Form Lyophilized Powder
Synonym Ins(2,4,5)-P3 sodium salt; 2,4,5-IP3 sodium salt

Specifications

Chemical Name or Material D-myo-Inositol-2,4,5-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD05864256
Synonym Ins(2,4,5)-P3 sodium salt; 2,4,5-IP3 sodium salt
Accessories

-30° to -10°C