missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,3,5-trisphosphate ammonium salt, 98%

Manufacturer:  Alfa Aesar™ J67016EXZ

 View more versions of this product

Catalog No. AAJ67016EXZ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

D-myo-Inositol-2,3,5-trisphosphate ammonium salt
0.1mg
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt
Lyophilized Powder
White
MFCD11114547
471.19
98%

Chemical Identifiers

MFCD11114547
Ins(2,3,5)-P3 ammonium salt; 2,3,5-IP3 ammonium salt
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents