missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt, 98%

Chemical Identifiers

MDL Number MFCD18427943
Synonym 2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67178EXZ
View Documents
Alfa Aesar™
J67178EXZ
0.1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67178EXD
View Documents
Alfa Aesar™
J67178EXD
0.5mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

MFCD18427943
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt

Specifications

White
MFCD18427943
588
98%
0.1mg
2,3,5,6-IP4 sodium salt; Ins(2,3,5,6)P4 sodium salt
Lyophilized Powder
D-myo-Inositol-2,3,5,6-tetrakis(phosphate) sodium salt
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C