Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-2,3,4,5-tetrakis(phosphate) ammonium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-2,3,4,5-tetrakis(phosphate) ammonium salt
Formula Weight 568.2
MDL Number MFCD11978039
Synonym Ins(2,3,4,5)-P4 ammonium salt; 2,3,4,5-IP4 ammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67024EXD

 
alfa aesar™
J67024EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67024EXZ

 
alfa aesar™
J67024EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 568.2
Chemical Name or Material D-myo-Inositol-2,3,4,5-tetrakis(phosphate) ammonium salt
Assay 98%
MDL Number MFCD11978039
Synonym Ins(2,3,4,5)-P4 ammonium salt; 2,3,4,5-IP4 ammonium salt
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Physical Form Lyophilized Powder
Safety and Handling

-30° to -10°C