Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1-phosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1-phosphate sodium salt
Formula Weight 282.12
MDL Number MFCD12912248
Synonym Ins(1)P1 sodium salt; 1-IP1 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67038EXD

 
alfa aesar™
J67038EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67038EXZ

 
alfa aesar™
J67038EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67038MCR

 
alfa aesar™
J67038MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 282.12
MDL Number MFCD12912248
Chemical Name or Material D-myo-Inositol-1-phosphate sodium salt
Physical Form Lyophilized Powder
Assay 98%
Synonym Ins(1)P1 sodium salt; 1-IP1 sodium salt
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C