Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1-phosphate cyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1-phosphate cyclohexylammonium salt
Formula Weight 458.48
MDL Number MFCD00161485
Synonym Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67121EXD

  SDS
alfa aesar™
J67121EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67121LB0

  SDS
alfa aesar™
J67121LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67121MCR

  SDS
alfa aesar™
J67121MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 458.48
MDL Number MFCD00161485
Chemical Name or Material D-myo-Inositol-1-phosphate cyclohexylammonium salt
Physical Form Lyophilized Powder
Assay 95%
Synonym Ins(1)P1 cyclohexylammonium salt; 1-IP1 cyclohexyl ammonium salt
Solubility Information Soluble in water
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C