Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,5,6-trisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,5,6-trisphosphate sodium salt
CAS 120965-76-6
Formula Weight 486.04
MDL Number MFCD12912676
Synonym Ins(1,5,6)-P3 sodium salt; 1,5,6-IP3 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67076EXD

 
alfa aesar™
J67076EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67076EXZ

 
alfa aesar™
J67076EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-1,5,6-trisphosphate sodium salt
Physical Form Lyophilized Powder
MDL Number MFCD12912676
CAS 120965-76-6
Synonym Ins(1,5,6)-P3 sodium salt; 1,5,6-IP3 sodium salt
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Assay 98%
Color White to off-white
Safety and Handling

-30° to -10°C