Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,4,5-trisphosphate sodium salt, 98%


Quantity 0.1mg

Manufacturer: alfa aesar™  J67063EXZ

 View more versions of this product

Catalog No. AAJ67063EXZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-1,4,5-trisphosphate sodium salt
Assay 98%
Physical Form Lyophilized Powder
MDL Number MFCD00055051
Synonym Ins(1,4,5)P3 sodium salt; 1,4,5-IP3 sodium salt
Storage Note 1 -30° to -10°C
Quantity 0.1mg
Color White to off-white

Specifications

Chemical Name or Material D-myo-Inositol-1,4,5-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD00055051
Synonym Ins(1,4,5)P3 sodium salt; 1,4,5-IP3 sodium salt
Accessories

-30° to -10°C