Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,3-bisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,3-bisphosphate sodium salt
CAS 208584-52-5
Formula Weight 386.09
MDL Number MFCD03095587
Synonym Ins(1,3)-P2; 1,3-IP2 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66093EXD

  SDS
alfa aesar™
J66093EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66093EXZ

  SDS
alfa aesar™
J66093EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66093MCR

  SDS
alfa aesar™
J66093MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.09
Assay 98%
MDL Number MFCD03095587
Color White to off-white
Storage Note 1 -30° to -10°C
Chemical Name or Material D-myo-Inositol-1,3-bisphosphate sodium salt
Physical Form Lyophilized Powder
CAS 208584-52-5
Synonym Ins(1,3)-P2; 1,3-IP2 sodium salt
Quantity 0.5mg
Safety and Handling

-30° to -10°C