Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,3,5-trisphosphate sodium salt, 98%

Chemical Identifiers

Chemical Name or Material D-myo-Inositol-1,3,5-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD12912250
Synonym Ins(1,3,5)-P3; 1,3,5-IP3 sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66819EXD

  SDS
alfa aesar™
J66819EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66819EXZ

  SDS
alfa aesar™
J66819EXZ
0.1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66819MCR

  SDS
alfa aesar™
J66819MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.04
MDL Number MFCD12912250
Chemical Name or Material D-myo-Inositol-1,3,5-trisphosphate sodium salt
Physical Form Lyophilized Powder
Assay 98%
Synonym Ins(1,3,5)-P3; 1,3,5-IP3 sodium salt
Color White to off-white
Quantity 0.5mg
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C