Please login to your online account to display your discounted pricing

Alfa Aesar™ D-myo-Inositol-1,3,4-trisphosphate sodium salt, 98%


Quantity 0.5mg

Manufacturer: alfa aesar™  J66963EXD

 View more versions of this product

Catalog No. AAJ66963EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 486.04
Chemical Name or Material D-myo-Inositol-1,3,4-trisphosphate sodium salt
MDL Number MFCD03412019
Physical Form Lyophilized Powder
Assay 98%
Synonym Ins(1,3,4)-P3; 1,3,4-IP3 sodium salt
Quantity 0.5mg
Color White to off-white
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material D-myo-Inositol-1,3,4-trisphosphate sodium salt
Formula Weight 486.04
MDL Number MFCD03412019
Synonym Ins(1,3,4)-P3; 1,3,4-IP3 sodium salt
Accessories

-30° to -10°C